Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D and C Yellow No. 8, 85%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
518-47-8
C20H10Na2O5
376.28
NJDNXYGOVLYJHP-UHFFFAOYSA-L
disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
[Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12
518-47-8
C20H10Na2O5
376.28
NJDNXYGOVLYJHP-UHFFFAOYSA-L
disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
[Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12

D1146, 518-47-8, C20H10O5Na2

Spectrum™ D and C Yellow No. 8 , or Fluorescein Sodium, is a synthetic pigment sometimes referred to in general terms as a xanthene color. In cosmetics and personal care products, Yellow 8 is used in the formulation of bath products, bubble baths, cleansing products, depilatories, permanent waves, and shaving cream. Yellow 8 is subject to certification by the Food and Drug Administration (FDA). Yellow 8 is a straight color.
CAS Max % 1
Arsenic (As) 3 ppm
Percent Purity 85%
Lead (Pb) 20 ppm
Mercury (Hg) 1 ppm
Packaging Amber Glass Bottle
Quantity 25 g