Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D(+)-Carvone, 96%, natural, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
2244-16-8
C10H14O
150.22
MFCD00062997
ULDHMXUKGWMISQ-VIFPVBQESA-N
d-carvone,+-carvone,s-+-carvone,s-carvone,s-2-methyl-5-prop-1-en-2-yl cyclohex-2-enone,+-s-carvone,carvone, +,d +-carvone,carvol,s-+-p-mentha-6,8-dien-2-one
16724
CHEBI:15399
(5S)-2-methyl-5-prop-1-en-2-ylcyclohex-2-en-1-one
CC1=CCC(CC1=O)C(=C)C
2244-16-8
C10H14O
150.22
MFCD00062997
ULDHMXUKGWMISQ-VIFPVBQESA-N
d-carvone,+-carvone,s-+-carvone,s-carvone,s-2-methyl-5-prop-1-en-2-yl cyclohex-2-enone,+-s-carvone,carvone, +,d +-carvone,carvol,s-+-p-mentha-6,8-dien-2-one
16724
CHEBI:15399
(5S)-2-methyl-5-prop-1-en-2-ylcyclohex-2-en-1-one
CC1=CCC(CC1=O)C(=C)C
Specific Rotation + 57.00
Packaging Glass bottle
Boiling Point 230.0°C
Color Colorless to Yellow
Density 0.9560g/mL
Specific Gravity 0.956
Refractive Index 1.4950 to 1.5010
Flash Point 97°C
Infrared Spectrum Authentic
Quantity 100g
Assay Percent Range 96%
Beilstein 07, 153
Merck Index 14, 1874
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, ether, chloroform,, propylene glycol and mineral oils, insoluble in glycerol
Formula Weight 150.22
Physical Form Liquid
Percent Purity 96%
Chemical Name or Material D(+)-Carvone, 98%
Show More Show Less

EINECSNumber : 218-827-2

RTECSNumber : OS8670000

TSCA : TSCA

Recommended Storage : May darken on storage