Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
D(+)-Carvone, 96%, natural, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• D(+)-Carvone belongs to terpene family and is considered a monoterpene ketone naturally found mainly in Achillea ageratum and Curcuma pierreana
Application
• D(+)-Carvone has shown a suppressant effect against high-fat diet induced weight gain in mice
• It has been used as a flavoring agent
• It has been used in organic synthesis as starting material for the asymmetric total synthesis of natural products, including the synthesis of the terpenoid quassin
• In vitro studies demonstrated that this compound can inhibit cellulase-induced capsidiol production in Nicotiana tabacum by interfering with protein isoprenylation
Specifications
Color | Colorless to Yellow |
Density | 0.9560g/mL |
Boiling Point | 230.0°C |
Flash Point | 97°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 95% min. (GC) |
Packaging | Glass bottle |
Refractive Index | 1.4950 to 1.5010 |
Quantity | 25 g |
Beilstein | 07, 153 |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
EINECSNumber | 218-827-2 |
RTECSNumber | OS8670000 |
TSCA | TSCA |
Recommended Storage | May darken on storage |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction