Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ D-Cycloserine, 20mg/mL in distilled water, sterile-filtered

Click to view available options
Quantity:
25mL
50mL
68-41-7
C3H6N2O2
102.093
MFCD00005353
DYDCUQKUCUHJBH-UWTATZPHSA-N
d-cycloserine,cycloserine,seromycin,orientomycin,oxamycin,cyclorin,cyclo-d-serine,cicloserina,farmiserina,miroseryn
6234
CHEBI:40009
(4R)-4-amino-1,2-oxazolidin-3-one
C1C(C(=O)NO1)N
68-41-7
C3H6N2O2
102.093
MFCD00005353
DYDCUQKUCUHJBH-UWTATZPHSA-N
d-cycloserine,cycloserine,seromycin,orientomycin,oxamycin,cyclorin,cyclo-d-serine,cicloserina,farmiserina,miroseryn
6234
CHEBI:40009
(4R)-4-amino-1,2-oxazolidin-3-one
C1C(C(=O)NO1)N
Physical Form Liquid
Quantity 25mL
Chemical Name or Material D-Cycloserine, 20mg/mL in distilled water, sterile-filtered

RTECSNumber : NY2975000

storageNote1 : -30° to -10°C