Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Cycloserine, Calbiochem™

Click to view available options
5g; Glass bottle
68-41-7
C3H6N2O2
102.093
DYDCUQKUCUHJBH-UWTATZPHSA-N
d-cycloserine,cycloserine,seromycin,orientomycin,oxamycin,cyclorin,cyclo-d-serine,cicloserina,farmiserina,miroseryn
6234
CHEBI:40009
(4R)-4-amino-1,2-oxazolidin-3-one
C1C(C(=O)NO1)N
68-41-7
C3H6N2O2
102.093
DYDCUQKUCUHJBH-UWTATZPHSA-N
d-cycloserine,cycloserine,seromycin,orientomycin,oxamycin,cyclorin,cyclo-d-serine,cicloserina,farmiserina,miroseryn
6234
CHEBI:40009
(4R)-4-amino-1,2-oxazolidin-3-one
C1C(C(=O)NO1)N

Partial agonist of NMDA receptors at the glycine modulatory site. Reported to improve memory-dependent tasks in rats.

  • Partial agonist of NMDA receptors at the glycine modulatory site
  • Reported to improve memory-dependent tasks in rats
  • Neutral and acidic solutions are unstable
Packaging Glass bottle
Purity ≥95% by HPLC
Physical Form White to pale yellow crystalline powder
Quantity 5g
Chemical Name or Material D-Cycloserine

RTECSNumber : NY2975000