Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ D-(+)-Dextrose, Anhydrous, Thermo Scientific™

Click to view available options
Quantity:
5 lb.
25 lb.
100 lb.
Packaging:
Plastic bottle
50-99-7
C6H12O6
180.156
MFCD00063774
GZCGUPFRVQAUEE-SLPGGIOYSA-N
2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal,d +-glucose,aldehydo-d-glucose,glucose, anhydrous,dextrose, anhydrous,d-glucose, anhydrous,glucosteril,anhydrous dextrose,dextrose solution,d-glucose in linear form
107526
CHEBI:42758
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
C(C(C(C(C(C=O)O)O)O)O)O
50-99-7
C6H12O6
180.156
MFCD00063774
GZCGUPFRVQAUEE-SLPGGIOYSA-N
2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal,d +-glucose,aldehydo-d-glucose,glucose, anhydrous,dextrose, anhydrous,d-glucose, anhydrous,glucosteril,anhydrous dextrose,dextrose solution,d-glucose in linear form
107526
CHEBI:42758
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
C(C(C(C(C(C=O)O)O)O)O)O
Packaging Plastic bottle
Quantity 5 lb.
Melting Point 147°C to 153°C
Linear Formula CH2OH(CHOH)4CHO
Formula Weight 180.16
Physical Form Crystalline Powder
Chemical Name or Material D-(+)-Dextrose