Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ D-dimer Monoclonal Antibody (22)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ ABS0152202
Description
ABS 015-22-02 detects D-dimer from human and dog samples. ABS 015-22-02 has been successfully used in ELISA procedures. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.
Fibrinogen is the main protein of blood coagulation system. It consists of two identical subunits that contain three polypeptide chains: alpha, beta and gamma. The process of blood coagulation results in the activation of fibrinogen into fibrin by thrombin and fibrin polymerization. Fibrin clot is then digested by plasmin, and fibrin degradation products of different molecular weights are released into the bloodstream.
Specifications
D-dimer | |
Monoclonal | |
1.10 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P02671, P68213 | |
FGA | |
Purified human D-dimer. | |
400 μL | |
Primary | |
Human, Canine | |
Antibody | |
IgG1 κ |
ELISA, Western Blot | |
22 | |
Unconjugated | |
FGA | |
carboxy-terminal region; ENSMUSG00000059807; FGA; Fib; Fib2; fibrinogen A-alpha-chain; fibrinogen alpha chain; fibrinogen, A alpha polypeptide; Fibrinopeptide A; MGC119422; MGC119423; MGC119425 | |
Mouse | |
Protein A/G | |
RUO | |
2243, 475473 | |
4°C, store in dark | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction