Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
D-Dimer Monoclonal Antibody (OTI10A4), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene CF813880


Description
D-Dimer Monoclonal antibody specifically detects D-Dimer in Human samples. It is validated for ELISASpecifications
D-Dimer | |
Monoclonal | |
See Label | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P02671 | |
Mouse | |
Protein A/G | |
RUO | |
2243 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
ELISA | |
OTI10A4 | |
Unconjugated | |
FGA | |
carboxy-terminal region; ENSMUSG00000059807; FGA; Fib; Fib2; fibrinogen A-alpha-chain; fibrinogen alpha chain; fibrinogen, A alpha polypeptide; Fibrinopeptide A; MGC119422; MGC119423; MGC119425 | |
Native Human D-Dimer protein | |
100 μg | |
Primary | |
Human | |
Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction