Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-erythro-Sphingosine (synthetic), Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
This item is not returnable. View return policy
123-78-4
C18H37NO2
299.499
WWUZIQQURGPMPG-KRWOKUGFSA-N
sphingosine,d-erythro-sphingosine,4-sphingenine,d-sphingosine,sphing-4-enine,cerebroside,sphingenine,4-trans-sphingenine,trans-d-erythro-2-amino-4-octadecene-1,3-diol,2s,3r,4e-2-amino-4-octadecene-1,3-diol
5280335
CHEBI:16393
(E,2S,3R)-2-aminooctadec-4-ene-1,3-diol
CCCCCCCCCCCCCC=CC(C(CO)N)O
This item is not returnable. View return policy
123-78-4
C18H37NO2
299.499
WWUZIQQURGPMPG-KRWOKUGFSA-N
sphingosine,d-erythro-sphingosine,4-sphingenine,d-sphingosine,sphing-4-enine,cerebroside,sphingenine,4-trans-sphingenine,trans-d-erythro-2-amino-4-octadecene-1,3-diol,2s,3r,4e-2-amino-4-octadecene-1,3-diol
5280335
CHEBI:16393
(E,2S,3R)-2-aminooctadec-4-ene-1,3-diol
CCCCCCCCCCCCCC=CC(C(CO)N)O

Protein kinase C inhibitor

Inhibitor of protein kinase C and calmodulin-dependent enzymes, but may stimulate mast cells by activation of protein kinase C.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 299.5
Percent Purity >98%
Chemical Name or Material D-erythro-Sphingosine (synthetic)

Recommended Storage : Store at -20°C