Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D(+)-Fenchone, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
4695-62-9
C10H16O
152.24
MFCD00070689
LHXDLQBQYFFVNW-XCBNKYQSSA-N
+-fenchone,1s-1,3,3-trimethylbicyclo 2.2.1 heptan-2-one,1s-1,3,3-trimethyl-norbornan-2-one
1201521
CHEBI:165
(1R,4S)-2,2,4-trimethylbicyclo[2.2.1]heptan-3-one
CC1(C2CCC(C2)(C1=O)C)C
4695-62-9
C10H16O
152.24
MFCD00070689
LHXDLQBQYFFVNW-XCBNKYQSSA-N
+-fenchone,1s-1,3,3-trimethylbicyclo 2.2.1 heptan-2-one,1s-1,3,3-trimethyl-norbornan-2-one
1201521
CHEBI:165
(1R,4S)-2,2,4-trimethylbicyclo[2.2.1]heptan-3-one
CC1(C2CCC(C2)(C1=O)C)C
Melting Point 5°C to 7°C
Boiling Point 63°C to 65°C (13.0mmHg)
Flash Point 62°C
Specific Gravity 0.95
Refractive Index 1.4610 to 1.464
Specific Rotation + 59
Color Colorless to Yellow
Specific Rotation Condition +59.00 (20.00°C neat)
Packaging Glass bottle
Density 0.9500g/mL
Infrared Spectrum Authentic
Quantity 100g
Assay Percent Range 96% min. (GC)
Beilstein 07, IV, 212
Merck Index 15, 4000
Solubility Information Solubility in water: practically insoluble in water. Other solubilities: soluble in ethanol,ether,common organic solvents
Formula Weight 152.24
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material D(+)-Fenchone, 97%
Show More Show Less

EINECSNumber : 225-160-