SHOWA™ D-Flex™ Gloves

Ideal for light-duty applications

Manufacturer: SHOWA™ 910C09

 View more versions of this product

Catalog No. 19-048-577D

  Check Availability
Add to Cart

    Specifications

    Glove Cuff Style Knit
    For Use With (Application) machine or compartment assembly, bottling, sheet Metal handling, construction, recycling, Glass handling, razor blades
    Weight Light weight
    Coating Material Dotted textured
    Color White
    Cut Resistance Rating Level 4
    Hand Ambidextrous
    Material Polyethylene
    Size 9
    Style Dotted

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit