Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(-)-Fructose, BAKER ANALYZED™ Biochemical Reagent, J.T.Baker™

Click to view available options
Quantity:
100g
Packaging:
Poly Bottle
57-48-7
C6H12O6
180.156
BJHIKXHVCXFQLS-UYFOZJQFSA-N
d---fructose,d--fructose,3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one,furucton,arabino-hexulose,keto-d-fructose,d-levulose,sugar, fruit,fructose, d,krystar 300
5984
CHEBI:48095
(3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
C(C(C(C(C(=O)CO)O)O)O)O
57-48-7
C6H12O6
180.156
BJHIKXHVCXFQLS-UYFOZJQFSA-N
d---fructose,d--fructose,3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one,furucton,arabino-hexulose,keto-d-fructose,d-levulose,sugar, fruit,fructose, d,krystar 300
5984
CHEBI:48095
(3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
C(C(C(C(C(=O)CO)O)O)O)O

High quality biochemicals for research use

Packaging Poly Bottle
Quantity 100g
Formula Weight 180.16
Grade BAKER ANALYZED™ Biochemical Reagent
Chemical Name or Material D-(-)-Fructose