Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D(+)-Galactose, 99+%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
1kg
5kg
25kg
59-23-4
C6H12O6
MFCD00063833
59-23-4
C6H12O6
MFCD00063833
Vapor Pressure Negligible
Water 0.3% max. (K.F.)
Sulfated Ash 0.1% max. (on dry substance)
Specific Rotation + 79.75
Infrared Spectrum Authentic
Appearance (10% aq. soln.) clear, colorless
Color White
Melting Point 165.0°C to 168.0°C
Quantity 25kg
Assay Percent Range 99+%
Beilstein 31, 295
Merck Index 15, 4365
Solubility Information Solubility in water: 680g/L (25°C). Other solubilities: soluble in pyridine, slighlty soluble in alcohol
Formula Weight 180.16
Physical Form Powder
Percent Purity ≥99%
Chemical Name or Material D(+)-Galactose
Show More Show Less