Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(+)-Galacturonic Acid, Monohydrate, Approx. 80%, Spectrum™ Chemical
SDP

Catalog Number 18601997
Click to view available options
Quantity:
5 g
Packaging:
Amber Glass Bottle
91510-62-2
C6H9O7
0.00
AEMOLEFTQBMNLQ-DTEWXJGMSA-M
(2S,3R,4S,5R,6R)-3,4,5,6-tetrahydroxyoxane-2-carboxylate
*
91510-62-2
C6H9O7
0.00
AEMOLEFTQBMNLQ-DTEWXJGMSA-M
(2S,3R,4S,5R,6R)-3,4,5,6-tetrahydroxyoxane-2-carboxylate
*

G3072, 91510-62-2, C6H10O7•H2O

Spectrum™ Chemical D-(+)-Galacturonic Acid, Monohydrate , also known simply as D-Galacturonic acid, is an oxidized form of D-galactose and a sugar acid. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 5 g
Percent Purity Approx. 80%
Grade Reagent
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit