Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Gluconic Acid, 50% Aqueous Solution, Spectrum™

Click to view available options
Quantity:
1 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
526-95-4,7732-18-5
526-95-4,7732-18-5
Grade Reagent
Packaging Amber Glass Bottle
Quantity 4 L

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.