Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
D-Gluconic acid, calcium salt, 99%, ACROS Organics™
Click to view available options
Quantity:
250g
1kg
5kg
Packaging:
Plastic bottle
Plastic drum
Specifications
Additional Information | Organic impurities and boric acid:Passes Test (EP) |
Alkalis and Magnesium | Magnesium and alkali metals: < 0.4% |
Assay Percent Range | 99% |
Linear Formula | (HOCH2[CH(OH)]4CO2)2Ca |
Chloride | 700ppm max. |
Beilstein | 03, 544 |
Merck Index | 15, 1671 |
Formula Weight | 430.38 |
Physical Form | Crystals or Crystalline Powder |
Percent Purity | 99% |
Show More |