Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Gluconic acid, calcium salt, 99%, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
5kg
Packaging:
Plastic bottle
Plastic drum
299-28-5
C12H22CaO14
430.372
MFCD00064209
NEEHYRZPVYRGPP-IYEMJOQQSA-L
calcium gluconate,calcium d-gluconate,calciofon,calglucon,glucobiogen,ebucin,calcicol,calcipur,calglucol,dragocal
9290
calcium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
C(C(C(C(C(C(=O)[O-])O)O)O)O)O.C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Ca+2]
299-28-5
C12H22CaO14
430.372
MFCD00064209
NEEHYRZPVYRGPP-IYEMJOQQSA-L
calcium gluconate,calcium d-gluconate,calciofon,calglucon,glucobiogen,ebucin,calcicol,calcipur,calglucol,dragocal
9290
calcium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
C(C(C(C(C(C(=O)[O-])O)O)O)O)O.C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Ca+2]
Additional Information Organic impurities and boric acid:Passes Test (EP)
Alkalis and Magnesium Magnesium and alkali metals: < 0.4%
Assay Percent Range 99%
Linear Formula (HOCH2[CH(OH)]4CO2)2Ca
Chloride 700ppm max.
Beilstein 03, 544
Merck Index 15, 1671
Formula Weight 430.38
Physical Form Crystals or Crystalline Powder
Percent Purity 99%
Heavy Metals 20ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Loss on Drying 2.0 % max. (1 g, 105°C, 16 hrs)
Packaging Plastic bottle
Reducing Sugars Sucrose and reducing sugars: < 1.0%
Specific Rotation + 10.20
Sulfate 500ppm max.
Specific Rotation Condition + 10.20 (22.00°C c=1,H2O)
Color White
Quantity 250g
Chemical Name or Material D-Gluconic acid, calcium salt
Show More Show Less