Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Glucosamine hydrochloride, 98+%, ACROS Organics™

Click to view available options
Quantity:
10g
100g
500g
2.5kg
Packaging:
Glass Bottle
Plastic bottle
66-84-2
C6H13NO5·HCl
66-84-2
C6H13NO5·HCl
Loss on Drying 1.0 % max. (105°C, 2 hrs)
Infrared Spectrum Authentic
Sulfated Ash 0.1 % max.
Packaging Plastic bottle
Quantity 500g
Color Brown-Yellow to Yellow
Specific Rotation Condition + 72.50 (20.00°C c=2,H2O,5hrs.)
Specific Rotation +70.0° to +73.0° (20°C, 589nm) (c=2.5, H2O) (after 3 hrs)
Assay Percent Range 98+%
Formula Weight 215.64
Physical Form Solid powder
Percent Purity 98.0 to 102.0% (on dry substance) (Total base)
Chemical Name or Material D-Glucosamine hydrochloride
Show More Show Less