Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Glucose-6-phosphate disodium salt dihydrate, Affymetrix/USB™

Click to view available options
Quantity:
5g
10g
100g
250g
Packaging:
Plastic bottle
3671-99-6
C6H11Na2O9P
304.098
MFCD00136037
VQLXCAHGUGIEEL-FAOVPRGRSA-L
d-glucose-6-phosphate disodium salt,sodium 2r,3r,4s,5r-2,3,4,5-tetrahydroxy-6-oxohexyl phosphate,sodium glucose 6-phosphate,disodium d-glucose 6-phosphate,glucose 6-phosphate sodium salt,glucose-6-phosphate disodium,disodium d-glucose-6-phosphate,d-glucose 6-disodium phosphate,d-glucose 6-phosphoric acid disodium salt,disodium 2r,3s,4r,5r-2,3,4,5-tetrahydroxy-6-phosphonooxy hexanal
18594434
disodium;[(2R,3R,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexyl] phosphate
C(C(C(C(C(C=O)O)O)O)O)OP(=O)([O-])[O-].[Na+].[Na+]
3671-99-6
C6H11Na2O9P
304.098
MFCD00136037
VQLXCAHGUGIEEL-FAOVPRGRSA-L
d-glucose-6-phosphate disodium salt,sodium 2r,3r,4s,5r-2,3,4,5-tetrahydroxy-6-oxohexyl phosphate,sodium glucose 6-phosphate,disodium d-glucose 6-phosphate,glucose 6-phosphate sodium salt,glucose-6-phosphate disodium,disodium d-glucose-6-phosphate,d-glucose 6-disodium phosphate,d-glucose 6-phosphoric acid disodium salt,disodium 2r,3s,4r,5r-2,3,4,5-tetrahydroxy-6-phosphonooxy hexanal
18594434
disodium;[(2R,3R,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexyl] phosphate
C(C(C(C(C(C=O)O)O)O)O)OP(=O)([O-])[O-].[Na+].[Na+]
Formula Weight 340.13
Physical Form Powder
Packaging Plastic bottle
Quantity 100g
Chemical Name or Material D-Glucose-6-phosphate disodium salt dihydrate