Learn More
R Biopharm Inc D-GLUCOSE TEST KIT
Manufacturer:  R Biopharm Inc 10716251035
Encompass
Intended use: UV-method for the determination of D-glucose in foodstuffs and other materials. Test-Combination for 3 X 45 determinations.
Catalog No. NC9508588