Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Glucurono-3,6-lactone, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
32449-92-6
32449-92-6
Packaging Amber Glass Bottle
Quantity 25 g
Appearance White Powder
CAS Max % 1
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.