Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ D-Glutamic acid 5-tert-butyl ester, 95%

Click to view available options
Quantity:
1g
5g
25g
45125-00-6
C9H17NO4
203.238
MFCD00153437
OIOAKXPMBIZAHL-ZCFIWIBFSA-N
h-d-glu otbu-oh,r-2-amino-5-tert-butoxy-5-oxopentanoic acid,d-glutamicacid, 5-1,1-dimethylethyl ester,2r-2-amino-5-tert-butoxy-5-oxopentanoic acid,ambotzhaa1231,l-glutamicacid5-tert-butylester,r-2-amino-5-tert-butoxy-5-oxopentanoicacid
7016383
(2R)-2-amino-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid
CC(C)(C)OC(=O)CCC(C(=O)O)N
45125-00-6
C9H17NO4
203.238
MFCD00153437
OIOAKXPMBIZAHL-ZCFIWIBFSA-N
h-d-glu otbu-oh,r-2-amino-5-tert-butoxy-5-oxopentanoic acid,d-glutamicacid, 5-1,1-dimethylethyl ester,2r-2-amino-5-tert-butoxy-5-oxopentanoic acid,ambotzhaa1231,l-glutamicacid5-tert-butylester,r-2-amino-5-tert-butoxy-5-oxopentanoicacid
7016383
(2R)-2-amino-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid
CC(C)(C)OC(=O)CCC(C(=O)O)N
Applications
It is employed as a intermediate for pharmaceutical and organic synthesis.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Solubility Information Slightly soluble in water.
Formula Weight 203.24
Physical Form Solid
Percent Purity 95%
Quantity 5g
Chemical Name or Material D-Glutamic acid 5-tert-butyl ester

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold