Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D(-)-Gulonic acid γ-lactone, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
6322-07-2
98.5% min. (HPLC)
C6H10O6
MFCD00005391
D(-)-Gulonic acid γ-lactone, 99%
6322-07-2
98.5% min. (HPLC)
C6H10O6
MFCD00005391
D(-)-Gulonic acid γ-lactone, 99%
Melting Point 182°C to 188°C
Specific Rotation -54.5
Color White
Packaging Glass bottle
Specific Rotation Condition −54.50° (20°C c=4,water)
Quantity 25g
Loss on Drying 1% max.
Infrared Spectrum Authentic
Beilstein 18, 204
InChI Key SXZYCXMUPBBULW-UHFFFAOYNA-N
SMILES OCC(O)C1OC(=O)C(O)C1O
IUPAC Name 5-(1,2-dihydroxyethyl)-3,4-dihydroxyoxolan-2-one
Molecular Weight (g/mol) 178.14
Formula Weight 178.14
Physical Form Powder or Cyrstals
Percent Purity 99%
Show More Show Less

EINECSNumber : 228-678-5