Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ D-Histidine methyl ester dihydrochloride, 95%

Click to view available options
Quantity:
1g
5g
25g
4467-54-3
C7H13Cl2N3O2
242.1
MFCD00066137
DWAYENIPKPKKMV-QYCVXMPOSA-N
d-histidine methyl ester dihydrochloride,r-methyl 2-amino-3-1h-imidazol-4-yl propanoate dihydrochloride,h-d-his-ome 2hcl,h-d-his-ome.2hcl,d-histidine methylester dihydrochloride,c7h11n3o2.2hcl,methyl 2r-2-amino-3-1h-imidazol-4-yl propanoate dihydrochloride,d-histidine methyl ester di hydrochloride,d-his-ome.2hcl,d-histidinemethylesterdihydrochloride
12658398
methyl (2R)-2-amino-3-(1H-imidazol-5-yl)propanoate;dihydrochloride
COC(=O)C(CC1=CN=CN1)N.Cl.Cl
4467-54-3
C7H13Cl2N3O2
242.1
MFCD00066137
DWAYENIPKPKKMV-QYCVXMPOSA-N
d-histidine methyl ester dihydrochloride,r-methyl 2-amino-3-1h-imidazol-4-yl propanoate dihydrochloride,h-d-his-ome 2hcl,h-d-his-ome.2hcl,d-histidine methylester dihydrochloride,c7h11n3o2.2hcl,methyl 2r-2-amino-3-1h-imidazol-4-yl propanoate dihydrochloride,d-histidine methyl ester di hydrochloride,d-his-ome.2hcl,d-histidinemethylesterdihydrochloride
12658398
methyl (2R)-2-amino-3-(1H-imidazol-5-yl)propanoate;dihydrochloride
COC(=O)C(CC1=CN=CN1)N.Cl.Cl
Beilstein 6491805
Formula Weight 242.1
Physical Form Solid
Percent Purity 95%
Color White
Quantity 5g
Sensitivity Hygroscopic
Chemical Name or Material D-Histidine methyl ester dihydrochloride

storageNote1 : 2° to 8°C