Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ D-(+)-Isoascorbic acid sodium salt monohydrate, 99%

Manufacturer:  Alfa Aesar™ H336700E

Quantity 2500g

 View more versions of this product

Catalog No. AAH336700E


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

63524-04-9
216.12 (198.11 Anhydrous)
Sodium erythorbate monohydrate; D-erythro-Hex-2-enonic acid gamma-lactone monosodium salt monohydrate
C6H7NaO6·H2O
MFCD03788863
D-(+)-Isoascorbic acid sodium salt monohydrate

Specifications

+97° (c=10 in Water)
63524-04-9
216.12 (198.11 Anhydrous)
Sodium erythorbate monohydrate; D-erythro-Hex-2-enonic acid gamma-lactone monosodium salt monohydrate
99%
2500g
C6H7NaO6·H2O
MFCD03788863
D-(+)-Isoascorbic acid sodium salt monohydrate

Safety and Handling

SDS & Safety Documents

RTECSNumber : MP8910000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit