Alfa Aesar™ D-(+)-Isoascorbic acid sodium salt monohydrate, 99%

Manufacturer: Alfa Aesar™ H3367036

Quantity 500g

 View more versions of this product

Catalog No. AAH3367036

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 63524-04-9
    Molecular Formula C6H7NaO6·H2O
    Formula Weight 216.12 (198.11 Anhydrous)
    MDL Number MFCD03788863
    Synonym Sodium erythorbate monohydrate; D-erythro-Hex-2-enonic acid gamma-lactone monosodium salt monohydrate
    Chemical Name or Material D-(+)-Isoascorbic acid sodium salt monohydrate

    Specifications

    Formula Weight 216.12 (198.11 Anhydrous)
    TSCA TSCA
    Molecular Formula C6H7NaO6·H2O
    MDL Number MFCD03788863
    CAS 63524-04-9
    Synonym Sodium erythorbate monohydrate; D-erythro-Hex-2-enonic acid gamma-lactone monosodium salt monohydrate
    RTECS Number MP8910000
    Optical Rotation +97° (c=10 in Water)
    Quantity 500g
    Chemical Name or Material D-(+)-Isoascorbic acid sodium salt monohydrate
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit