Learn More
BioVision IncSupplier Diversity Partner D-Lactate Colorimetric Assay Kit: Colorimetric Assay to Measure D-Lactate in a variety of Biological Samples such as Cells, Tissues etc. within 40 min. Rapid, Simple & Sensitive. Detection Range: 0.01 mM – 10 mM. 100 Assays DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K667100
D-Lactate production in mammals, mainly due to the glyoxalase pathway, is extremely low, with normal serum concentrations in the nano to micromolar range. Typically, elevated D-lactate levels which can rise to millimolar levels, are due to bacterial infection or short bowel syndrome in humans. Abnormally high concentrations of D-lactate are considered indicative of sepsis, ischemia or trauma. Due to slow metabolism and excretion, high D-lactate can cause acidosis and encephalopathy. BioVision’s D-Lactate Assay Kit provides a fast, easy way to accurately measure D-lactate in a variety of biological samples. In the D-Lactate Assay Kit, D-lactate is specifically oxidized by D-lactate dehydrogenase and generates proportional color (λmax = 450 nm). The kit detects D-Lactate in samples such as serum, plasma, cells, culture and fermentation media. The useful concentration range in samples is 0.01 mM – 10 mM D-lactate. BioVision charges an Ice pack fee for each order.
Catalog No. 50-111-9826