Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-D-Lactose monohydrate, Acros Organics

Click to view available options
Quantity:
250g
1kg
Packaging:
Plastic bottle
5989-81-1
C12H24O12
360.312
WSVLPVUVIUVCRA-KPKNDVKVSA-N
alpha-d-lactose monohydrate,alpha-lactose monohydrate,lactose monohydrate,lactose, monohydrate,unii-ewq57q8i5x,respitose,alpha-lactose hydrate,a-lactose monohydrate,lactose monohydrate nf,ewq57q8i5x
104938
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol;hydrate
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O.O
5989-81-1
C12H24O12
360.312
WSVLPVUVIUVCRA-KPKNDVKVSA-N
alpha-d-lactose monohydrate,alpha-lactose monohydrate,lactose monohydrate,lactose, monohydrate,unii-ewq57q8i5x,respitose,alpha-lactose hydrate,a-lactose monohydrate,lactose monohydrate nf,ewq57q8i5x
104938
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol;hydrate
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O.O
Quantity 250g
Packaging Plastic bottle
Formula Weight 360.3
Chemical Name or Material α-D-Lactose monohydrate