Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ D-Leucine ethyl ester hydrochloride, 98%

Click to view available options
Quantity:
1g
5g
25g
73913-65-2
C8H18ClNO2
195.687
MFCD00190936
NOUDPBCEONUCOV-OGFXRTJISA-N
d-leucine ethyl ester hydrochloride,ethyl d-leucinate hydrochloride,h-d-leu-oet.hcl,r-ethyl 2-amino-4-methylpentanoate hydrochloride,ethyl 2r-2-amino-4-methylpentanoate hydrochloride,h-d-leu-oet?cl,h-d-leu-oet. hcl,h-d-leu-oet hydrochloride,ethyld-leucinatehydrochloride,d-leucine ethylester hydrochloride
11600850
ethyl (2R)-2-amino-4-methylpentanoate;hydrochloride
CCOC(=O)C(CC(C)C)N.Cl
73913-65-2
C8H18ClNO2
195.687
MFCD00190936
NOUDPBCEONUCOV-OGFXRTJISA-N
d-leucine ethyl ester hydrochloride,ethyl d-leucinate hydrochloride,h-d-leu-oet.hcl,r-ethyl 2-amino-4-methylpentanoate hydrochloride,ethyl 2r-2-amino-4-methylpentanoate hydrochloride,h-d-leu-oet?cl,h-d-leu-oet. hcl,h-d-leu-oet hydrochloride,ethyld-leucinatehydrochloride,d-leucine ethylester hydrochloride
11600850
ethyl (2R)-2-amino-4-methylpentanoate;hydrochloride
CCOC(=O)C(CC(C)C)N.Cl
Formula Weight 195.69
Physical Form Powder
Percent Purity 98%
Color White
Melting Point 121°C to 127°C
Quantity 5g
Sensitivity Hygroscopic
Chemical Name or Material D-Leucine ethyl ester hydrochloride

storageNote1 : 2° to 8°C