Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ D-Leucine methyl ester hydrochloride, 98%

Click to view available options
Quantity:
5g
25g
100g
5845-53-4
C7H16ClNO2
181.66
MFCD00066115
DODCBMODXGJOKD-FYZOBXCZSA-N
d-leucine methyl ester hydrochloride,r-methyl 2-amino-4-methylpentanoate hydrochloride,h-d-leu-ome.hcl,d-leucine, methyl ester, hydrochloride,methyl 2r-2-amino-4-methylpentanoate, chloride,h-d-leu-ome hcl,methyl d-leucinate hydrochloride,d-leu-och3.hcl,d-leucine,methyl ester,pubchem10893
11600782
methyl (2R)-2-amino-4-methylpentanoate;hydrochloride
CC(C)CC(C(=O)OC)N.Cl
5845-53-4
C7H16ClNO2
181.66
MFCD00066115
DODCBMODXGJOKD-FYZOBXCZSA-N
d-leucine methyl ester hydrochloride,r-methyl 2-amino-4-methylpentanoate hydrochloride,h-d-leu-ome.hcl,d-leucine, methyl ester, hydrochloride,methyl 2r-2-amino-4-methylpentanoate, chloride,h-d-leu-ome hcl,methyl d-leucinate hydrochloride,d-leu-och3.hcl,d-leucine,methyl ester,pubchem10893
11600782
methyl (2R)-2-amino-4-methylpentanoate;hydrochloride
CC(C)CC(C(=O)OC)N.Cl
Beilstein 3655228
Formula Weight 181.66
Physical Form Solid
Percent Purity 98%
Color White
Melting Point 151°C
Quantity 25g
Sensitivity Hygroscopic
Chemical Name or Material D-Leucine methyl ester hydrochloride