Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D(-)-Luciferin, ACROS Organics™

Click to view available options
Quantity:
10mg
50mg
Packaging:
Glass bottle
2591-17-5
C11H8N2O3S2
280.316
MFCD00042929
IWJYWBVPCGUPLO-SSDOTTSWSA-N
luciferin,d-luciferin,s-2-6-hydroxybenzo d thiazol-2-yl-4,5-dihydrothiazole-4-carboxylic acid,luciferin firefly,s-2-6-hydroxy-2-benzothiazolyl-2-thiazoline-4-carboxylic acid,4s-2-6-oxo-1,3-benzothiazol-2-ylidene-1,3-thiazolidine-4-carboxylic acid,4-thiazolecarboxylicacid, 4,5-dihydro-2-6-hydroxy-2-benzothiazolyl-, 4s
7023066
(4S)-2-(6-oxo-1,3-benzothiazol-2-ylidene)-1,3-thiazolidine-4-carboxylic acid
C1C(NC(=C2N=C3C=CC(=O)C=C3S2)S1)C(=O)O
2591-17-5
C11H8N2O3S2
280.316
MFCD00042929
IWJYWBVPCGUPLO-SSDOTTSWSA-N
luciferin,d-luciferin,s-2-6-hydroxybenzo d thiazol-2-yl-4,5-dihydrothiazole-4-carboxylic acid,luciferin firefly,s-2-6-hydroxy-2-benzothiazolyl-2-thiazoline-4-carboxylic acid,4s-2-6-oxo-1,3-benzothiazol-2-ylidene-1,3-thiazolidine-4-carboxylic acid,4-thiazolecarboxylicacid, 4,5-dihydro-2-6-hydroxy-2-benzothiazolyl-, 4s
7023066
(4S)-2-(6-oxo-1,3-benzothiazol-2-ylidene)-1,3-thiazolidine-4-carboxylic acid
C1C(NC(=C2N=C3C=CC(=O)C=C3S2)S1)C(=O)O
Quantity 10mg
Packaging Glass bottle
Infrared Spectrum Authentic
Color White to Yellow
Specific Rotation -32.0° to -36.0° (20°C, 589nm) (c=1, DMF)
Molar Absorption Coefficient 18200L/mol.cm min., 6900L/(mol.cm) min.
Water 2% Max. (K.F.)
Merck Index 15, 4117
Formula Weight 280.3
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material D(-)-Luciferin
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

LIGHT SENSITIVE

EINECSNumber : 219-981-3