Learn More
Aat BioquestSupplier Diversity Partner D-Luciferin, free acid *CAS#: 2591-17-5*  DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 12501
Luciferin is the most popular and versatile bioluminescent substrate. The firefly luciferase/luciferin bioluminescent system is found in the firefly (Photinus pyralis) and several other beetles.
Catalog No. 50-195-3385