Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(+)-Malic acid, 98+%, Thermo Scientific Chemicals

Catalog Number AAA1168803
Click to view available options
Quantity:
1 g
5 g
25 g
636-61-3
C4H6O5
134.09
MFCD00004245
BJEPYKJPYRNKOW-UHFFFAOYNA-N
d-+-malic acid, d-malic acid, d +-malic acid, r-2-hydroxysuccinic acid, r-malic acid, malic acid, d, 2r-2-hydroxybutanedioic acid, l +-malic acid, d-malate
92824
CHEBI:30796
(2R)-2-hydroxybutanedioic acid
OC(CC(O)=O)C(O)=O
636-61-3
C4H6O5
134.09
MFCD00004245
BJEPYKJPYRNKOW-UHFFFAOYNA-N
d-+-malic acid, d-malic acid, d +-malic acid, r-2-hydroxysuccinic acid, r-malic acid, malic acid, d, 2r-2-hydroxybutanedioic acid, l +-malic acid, d-malate
92824
CHEBI:30796
(2R)-2-hydroxybutanedioic acid
OC(CC(O)=O)C(O)=O

D-(+)-Malic acid used as an acidulant and flavoring agent, food additive. And it is also used in the place of the less sour citric acid in sour sweets.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(+)-Malic acid used as an acidulant and flavoring agent, food additive. And it is also used in the place of the less sour citric acid in sour sweets.

Solubility
Soluble in methanol, ethanol, acetone, ether.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY
Melting Point 100°C to 104°C
Density 1.6
Quantity 1 g
Beilstein 1723540
Merck Index 14,5707
Solubility Information Soluble in methanol,ethanol,acetone,ether.
Optical Rotation +28° (c=5.5 in pyridine)
Formula Weight 134.09
Percent Purity ≥98%
Chemical Name or Material D- (+)-Malic acid
Hazard Category H315-H318-H335
Hazard Statement GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
EINECSNumber 211-262-2
RTECSNumber ON7260000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit