Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ D-Methionine ethyl ester hydrochloride, 95%

Click to view available options
Quantity:
5g
25g
100g
7512-43-8
C7H16ClNO2S
213.72
MFCD00153451
KPWCQEUBJAIORR-FYZOBXCZSA-N
h-d-met-oet.hcl,h-d-met-oet hcl,d-methionine ethyl ester hydrochloride,r-ethyl 2-amino-4-methylthio butanoate hydrochloride,ethyl 2r-2-amino-4-methylsulfanyl butanoate hydrochloride,c7h15no2s.hcl,h-d-met-oethc,h-d-met-oet?cl,h-d-met-oet. hcl,d-methionine ethylester hydrochloride
44717125
ethyl (2R)-2-amino-4-methylsulfanylbutanoate;hydrochloride
CCOC(=O)C(CCSC)N.Cl
7512-43-8
C7H16ClNO2S
213.72
MFCD00153451
KPWCQEUBJAIORR-FYZOBXCZSA-N
h-d-met-oet.hcl,h-d-met-oet hcl,d-methionine ethyl ester hydrochloride,r-ethyl 2-amino-4-methylthio butanoate hydrochloride,ethyl 2r-2-amino-4-methylsulfanyl butanoate hydrochloride,c7h15no2s.hcl,h-d-met-oethc,h-d-met-oet?cl,h-d-met-oet. hcl,d-methionine ethylester hydrochloride
44717125
ethyl (2R)-2-amino-4-methylsulfanylbutanoate;hydrochloride
CCOC(=O)C(CCSC)N.Cl
Formula Weight 213.72
Physical Form Solid
Percent Purity 95%
Quantity 100g
Chemical Name or Material D-Methionine ethyl ester hydrochloride

storageNote1 : 2° to 8°C