Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D(+)-Methioninol, 97%, ACROS Organics™

Click to view available options
Quantity:
250mg
Packaging:
Glass Bottle
87206-44-8
C5H13NOS
135.225
MFCD00235938
MIQJGZAEWQQAPN-RXMQYKEDSA-N
d-methioninol,d +-methioninol,h-d-met-ol,d-+-methioninol,r-2-amino-4-methylthio butan-1-ol,r-2-amino-4-methylsulfanyl-butan-1-ol,2r-2-amino-4-methylsulfanyl butan-1-ol,1-butanol,2-amino-4-methylthio-, 2r,h-d-methioninol,r-methioninol
7000073
(2R)-2-amino-4-methylsulfanylbutan-1-ol
CSCCC(CO)N
87206-44-8
C5H13NOS
135.225
MFCD00235938
MIQJGZAEWQQAPN-RXMQYKEDSA-N
d-methioninol,d +-methioninol,h-d-met-ol,d-+-methioninol,r-2-amino-4-methylthio butan-1-ol,r-2-amino-4-methylsulfanyl-butan-1-ol,2r-2-amino-4-methylsulfanyl butan-1-ol,1-butanol,2-amino-4-methylthio-, 2r,h-d-methioninol,r-methioninol
7000073
(2R)-2-amino-4-methylsulfanylbutan-1-ol
CSCCC(CO)N
Packaging Glass Bottle
Quantity 250mg
Specific Rotation Condition + 11.00 (20.00°C c=1,EtOH)
Specific Gravity 1.095
Specific Rotation + 11.00
Infrared Spectrum Authentic
Flash Point 113°C
Melting Point 31.0°C to 36.0°C
Boiling Point 120.0°C to 122.0°C (1.0 mmHg)
Density 1.0950g/mL
Color White
Assay Percent Range 97%
Linear Formula CH3SCH2CH2CH(NH2)CH2OH
Formula Weight 135.22
Physical Form Waxy Low Melting Mass
Percent Purity 97%
Chemical Name or Material D(+)-Methioninol
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.