Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-myo-Inositol-1,2,3,6-tetrakis(phosphate) sodium salt, 98%

Click to view available options
Quantity:
0.1mg
Quantity 0.1mg
Color White
MDL Number MFCD12912298
Synonym 1,2,3,6-IP4 sodium salt; Ins(1,2,3,6)-P4 sodium salt
Formula Weight 588
Physical Form Lyophilized Powder
Percent Purity 98%
Chemical Name or Material D-myo-Inositol-1,2,3,6-tetrakis(phosphate) sodium salt

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C