Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ D-myo-Inositol-1,2-bisphosphate sodium salt, 98%

Click to view available options
Quantity:
0.1mg
0.5mg
1mg
208584-51-4
C6H14Na2O12P2
386.093
MFCD10567083
PHZDHEPQFXTBRQ-JBGYRJLXSA-N
d-myo-inositol-1,2-diphosphate sodium salt
131875843
sodium;[(1S,2R,3S,4S,5R,6R)-2,3,4,5-tetrahydroxy-6-phosphonooxycyclohexyl] dihydrogen phosphate
C1(C(C(C(C(C1O)OP(=O)(O)O)OP(=O)(O)O)O)O)O.[Na].[Na]
208584-51-4
C6H14Na2O12P2
386.093
MFCD10567083
PHZDHEPQFXTBRQ-JBGYRJLXSA-N
d-myo-inositol-1,2-diphosphate sodium salt
131875843
sodium;[(1S,2R,3S,4S,5R,6R)-2,3,4,5-tetrahydroxy-6-phosphonooxycyclohexyl] dihydrogen phosphate
C1(C(C(C(C(C1O)OP(=O)(O)O)OP(=O)(O)O)O)O)O.[Na].[Na]
Quantity 0.1mg
Color White
Formula Weight 384.08
Physical Form Lyophilized Powder
Percent Purity 98%
Chemical Name or Material D-myo-Inositol-1,2-bisphosphate sodium salt

storageNote1 : -30° to -10°C