Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ D-myo-Inositol-1,3,4,5-tetrakis(phosphate) octasodium salt, 98%

Click to view available options
Quantity:
0.1mg
0.5mg
210488-61-2
C6H8Na8O18P4
675.925
MFCD00153818
QYQMPPDYTHNACQ-QAVKKEKOSA-F
d-myo-inositol-1,3,4,5-tetraphosphate sodium salt,d-myo-inositol-1,3,4,5-tetraphosphate sodium salt,d-myo-inositol-1,3,4,5-tetrakisphosphate, octanatrium salt,d-myo-inositol 1,3,4,5-tetrakis phosphoric acid disodium salt
51346578
octasodium;[(1R,2S,4S,5S)-2,4-dihydroxy-3,5,6-triphosphonatooxycyclohexyl] phosphate
C1(C(C(C(C(C1OP(=O)([O-])[O-])OP(=O)([O-])[O-])OP(=O)([O-])[O-])O)OP(=O)([O-])[O-])O.[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+]
210488-61-2
C6H8Na8O18P4
675.925
MFCD00153818
QYQMPPDYTHNACQ-QAVKKEKOSA-F
d-myo-inositol-1,3,4,5-tetraphosphate sodium salt,d-myo-inositol-1,3,4,5-tetraphosphate sodium salt,d-myo-inositol-1,3,4,5-tetrakisphosphate, octanatrium salt,d-myo-inositol 1,3,4,5-tetrakis phosphoric acid disodium salt
51346578
octasodium;[(1R,2S,4S,5S)-2,4-dihydroxy-3,5,6-triphosphonatooxycyclohexyl] phosphate
C1(C(C(C(C(C1OP(=O)([O-])[O-])OP(=O)([O-])[O-])OP(=O)([O-])[O-])O)OP(=O)([O-])[O-])O.[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+]
Quantity 0.5mg
Color White
Formula Weight 675.93
Physical Form Lyophilized Powder
Percent Purity 98%
Chemical Name or Material D-myo-Inositol-1,3,4,5-tetrakis(phosphate) octasodium salt

storageNote1 : -30° to -10°C