Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ D-myo-Inositol-1,3,4,6-tetrakis(phosphate) ammonium salt, 95%

Click to view available options
Quantity:
0.1mg
0.5mg
142507-74-2
C6H19NO18P4
517.102
MFCD01459915
ANQVRWQIKKBLLK-ADHKWWJESA-N
myo-inositol 1,3,4,6-tetrakis-phosphate ammonium salt,1r,2s,3s,4s,5r,6r-2,5-dihydroxy-3,4,6-tris phosphonooxy cyclohexyl oxyphosphonic acid amine
71308569
azane;[(1S,3R,4R,6S)-2,5-dihydroxy-3,4,6-triphosphonooxycyclohexyl] dihydrogen phosphate
C1(C(C(C(C(C1OP(=O)(O)O)OP(=O)(O)O)O)OP(=O)(O)O)OP(=O)(O)O)O.N
142507-74-2
C6H19NO18P4
517.102
MFCD01459915
ANQVRWQIKKBLLK-ADHKWWJESA-N
myo-inositol 1,3,4,6-tetrakis-phosphate ammonium salt,1r,2s,3s,4s,5r,6r-2,5-dihydroxy-3,4,6-tris phosphonooxy cyclohexyl oxyphosphonic acid amine
71308569
azane;[(1S,3R,4R,6S)-2,5-dihydroxy-3,4,6-triphosphonooxycyclohexyl] dihydrogen phosphate
C1(C(C(C(C(C1OP(=O)(O)O)OP(=O)(O)O)O)OP(=O)(O)O)OP(=O)(O)O)O.N
Color White
Quantity 0.5mg
Formula Weight 568.2
Physical Form Lyophilized Powder
Percent Purity 95%
Chemical Name or Material D-myo-Inositol-1,3,4,6-tetrakis(phosphate) ammonium salt

storageNote1 : -30° to -10°C