Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ D-myo-Inositol-1,3,5-trisphosphate sodium salt, 98%

Click to view available options
Quantity:
0.1mg
0.5mg
1mg
MFCD12912250
Ins(1,3,5)-P3; 1,3,5-IP3 sodium salt
MFCD12912250
Ins(1,3,5)-P3; 1,3,5-IP3 sodium salt
Quantity 1mg
Color White
Formula Weight 486.04
Physical Form Lyophilized Powder
Percent Purity 98%
Chemical Name or Material D-myo-Inositol-1,3,5-trisphosphate sodium salt

storageNote1 : -30° to -10°C