Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ D-myo-Inositol-1,4,6-trisphosphate sodium salt, 98%

Click to view available options
Quantity:
0.1mg
0.5mg
MFCD12912289
Ins(1,4,6)P3 sodium salt; 1,4,6-IP3 sodium salt
MFCD12912289
Ins(1,4,6)P3 sodium salt; 1,4,6-IP3 sodium salt
Quantity 0.5mg
Color White
Formula Weight 486.04
Physical Form Lyophilized Powder
Percent Purity 98%
Chemical Name or Material D-myo-Inositol-1,4,6-trisphosphate sodium salt

storageNote1 : -30° to -10°C