Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ D-myo-Inositol-1,5,6-trisphosphate sodium salt, 98%

Click to view available options
Quantity:
0.1mg
0.5mg
120965-76-6
MFCD12912676
d-myo-inositol-1,5,6-triphosphate, sodium salt,1,5,6-ip3 sodium salt,trisodium 1r,2r,3s,4r,5r,6r-2,3-bis hydrogen phosphonatooxy-4,5,6-trihydroxycyclohexyl hydrogen phosphate
120965-76-6
MFCD12912676
d-myo-inositol-1,5,6-triphosphate, sodium salt,1,5,6-ip3 sodium salt,trisodium 1r,2r,3s,4r,5r,6r-2,3-bis hydrogen phosphonatooxy-4,5,6-trihydroxycyclohexyl hydrogen phosphate
Color White
Quantity 0.1mg
Formula Weight 486.04
Physical Form Lyophilized Powder
Percent Purity 98%
Chemical Name or Material D-myo-Inositol-1,5,6-trisphosphate sodium salt

storageNote1 : -30° to -10°C