Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ D-myo-Inositol-1-phosphate cyclohexylammonium salt, 95%

Click to view available options
Quantity:
0.5mg
1mg
5mg
MFCD00161485
Ins(1)P1 cyclohexylammonium salt; 1-IP1 cyclohexyl ammonium salt
MFCD00161485
Ins(1)P1 cyclohexylammonium salt; 1-IP1 cyclohexyl ammonium salt
Quantity 1mg
Color White
Solubility Information Soluble in water
Formula Weight 458.48
Physical Form Lyophilized Powder
Percent Purity 95%
Chemical Name or Material D-myo-Inositol-1-phosphate cyclohexylammonium salt

storageNote1 : -30° to -10°C