Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ D-myo-Inositol-2,3,5,6-tetrakis(phosphate) sodium salt, 98%
Click to view available options
Quantity:
0.1mg
0.5mg
Specifications
Color | White |
Quantity | 0.1mg |
Formula Weight | 588 |
Physical Form | Lyophilized Powder |
Percent Purity | 98% |
Chemical Name or Material | D-myo-Inositol-2,3,5,6-tetrakis(phosphate) sodium salt |
Safety and Handling
storageNote1 : -30° to -10°C