Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ D-myo-Inositol-2,3,5,6-tetrakis(phosphate) sodium salt, 98%

Click to view available options
Quantity:
0.1mg
0.5mg
MFCD18427943
2,3,5,6-IP4 sodium salt; Ins(2,3,5,6)P4 sodium salt
MFCD18427943
2,3,5,6-IP4 sodium salt; Ins(2,3,5,6)P4 sodium salt
Quantity 0.5mg
Color White
Formula Weight 588
Physical Form Lyophilized Powder
Percent Purity 98%
Chemical Name or Material D-myo-Inositol-2,3,5,6-tetrakis(phosphate) sodium salt

storageNote1 : -30° to -10°C