Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ D-myo-Inositol-2,3,5-trisphosphate ammonium salt, 98%

Click to view available options
Quantity:
0.1mg
0.5mg
MFCD11114547
Ins(2,3,5)-P3 ammonium salt; 2,3,5-IP3 ammonium salt
MFCD11114547
Ins(2,3,5)-P3 ammonium salt; 2,3,5-IP3 ammonium salt
Quantity 0.5mg
Color White
Formula Weight 471.19
Physical Form Lyophilized Powder
Percent Purity 98%
Chemical Name or Material D-myo-Inositol-2,3,5-trisphosphate ammonium salt

storageNote1 : -30° to -10°C