Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ D-myo-Inositol-2,4-bisphosphate sodium salt, 98%

Click to view available options
Quantity:
0.1mg
0.5mg
1mg
MFCD18427940
Ins(2,4)-P2 sodium salt; 2,4-IP2 sodium salt
MFCD18427940
Ins(2,4)-P2 sodium salt; 2,4-IP2 sodium salt
Quantity 0.5mg
Color White
Formula Weight 384.08
Physical Form Lyophilized Powder
Percent Purity 98%
Chemical Name or Material D-myo-Inositol-2,4-bisphosphate sodium salt

storageNote1 : -30° to -10°C