Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Pantethine, ≥90%, MilliporeSigma™ Supelco™

$99.70 - $333.50

Specifications

CAS 16816-67-4
Percent Purity ≥90%
Grade Analytical Standard
Linear Formula C22H42N4O8S2
Physical Form Neat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-3322
View Documents
MilliporeSigma Supelco
P21251G
1 g
Each for $99.70
Add to cart
 
11-101-3321
View Documents
MilliporeSigma Supelco
P21255G
5 g
Each for $333.50
Add to cart
 
Description

Description

Featured Industries: Vitamins, Nutraceuticals and Natural Products
Specifications

Specifications

16816-67-4
Analytical Standard
Neat
≥90%
C22H42N4O8S2
Safety and Handling

Safety and Handling

Recommended Storage : 2°C to 8°C

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit