Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Pantethine, 98%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
16816-67-4
C22H42N4O8S2
554.72
DJWYOLJPSHDSAL-ROUUACIJSA-N
(2R)-N-[2-({2-[(2-{3-[(2R)-2,4-dihydroxy-3,3-dimethylbutanamido]propanamido}ethyl)disulfanyl]ethyl}carbamoyl)ethyl]-2,4-dihydroxy-3,3-dimethylbutanamide
CC(C)(CO)[C@@H](O)C(=O)NCCC(=O)NCCSSCCNC(=O)CCNC(=O)[C@H](O)C(C)(C)CO
16816-67-4
C22H42N4O8S2
554.72
DJWYOLJPSHDSAL-ROUUACIJSA-N
(2R)-N-[2-({2-[(2-{3-[(2R)-2,4-dihydroxy-3,3-dimethylbutanamido]propanamido}ethyl)disulfanyl]ethyl}carbamoyl)ethyl]-2,4-dihydroxy-3,3-dimethylbutanamide
CC(C)(CO)[C@@H](O)C(=O)NCCC(=O)NCCSSCCNC(=O)CCNC(=O)[C@H](O)C(C)(C)CO

P1024, 16816-67-4, C22H42N4O8S2

Spectrum™ D-Pantethine , also known as Bis-pantethine, can be used to treat acne and improve the blood-cholesterol profile. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Packaging Amber Glass Bottle
Quantity 1 kg
Water 18-22%
CAS Max % 1
Percent Purity 98%