Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D(-)-Penicillamine, 99%, Acros Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
52-67-5
C5H11NO2S
149.208
MFCD00064302
VVNCNSJFMMFHPL-VKHMYHEASA-N
d-penicillamine,penicillamine,cuprimine,d---penicillamine,depen,3-mercapto-d-valine,cuprenil,d-penamine,--penicillamine,d-mercaptovaline
5852
CHEBI:7959
(2S)-2-amino-3-methyl-3-sulfanylbutanoic acid
CC(C)(C(C(=O)O)N)S
52-67-5
C5H11NO2S
149.208
MFCD00064302
VVNCNSJFMMFHPL-VKHMYHEASA-N
d-penicillamine,penicillamine,cuprimine,d---penicillamine,depen,3-mercapto-d-valine,cuprenil,d-penamine,--penicillamine,d-mercaptovaline
5852
CHEBI:7959
(2S)-2-amino-3-methyl-3-sulfanylbutanoic acid
CC(C)(C(C(=O)O)N)S
Infrared Spectrum Authentic
Color White
Appearance (10% aq. soln.) Passes Test
Specific Rotation − 65.00
Sulfated Ash 0.1% max.
Absorbance 0.07 max. (c=0.2, H2O, 1 cm cell) at 268nm
Packaging Glass bottle
Specific Rotation Condition −65° (20°C c=5,1M NaOH,on dry)
Quantity 5g
Melting Point 212.0°C
Loss on Drying 0.5% max. (1 g, 60°C, 670 Pa) (P2O5)
pH 4.5 to 5.5 (1% soln.)
Assay Percent Range 99%
Linear Formula (CH3)2C(SH)CH(NH2)CO2H
Merck Index 15, 7197
Formula Weight 149.21
Physical Form Powder
Percent Purity 99%
Chemical Name or Material D(-)-Penicillamine, 99+%
Show More Show Less

EINECSNumber : 200-148-8

RTECSNumber : YV9425000